Triacetin, 99%, Thermo Scientific Chemicals
Triacetin, 99%, Thermo Scientific Chemicals
Triacetin, 99%, Thermo Scientific Chemicals
Triacetin, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Triacetin, 99%, Thermo Scientific Chemicals
CAS: 102-76-1 | C9H14O6 | 218.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
500 mL
2.5 L
Packaging:
Glass bottle
Catalog number ACR139220010
Price (MYR)
759.00
Add to cart
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
759.00
Add to cart
Chemical Identifiers
CAS56-84-8
IUPAC Name2-aminobutanedioic acid
Molecular FormulaC4H7NO4
InChI KeyCKLJMWTZIZZHCS-UHFFFAOYNA-N
SMILESNC(CC(O)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Appearance (Color)White
Assay (Non-aqueous acid-base Titration)≥98.0 to ≤102.0% (dry wt. basis)
Loss on Drying≤0.5% (105°C)