o-Toluidine, 99%, Thermo Scientific Chemicals
o-Toluidine, 99%, Thermo Scientific Chemicals
o-Toluidine, 99%, Thermo Scientific Chemicals
o-Toluidine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
o-Toluidine, 99%, Thermo Scientific Chemicals
CAS: 95-53-4 | C7H9N | 107.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2.5 L
Packaging:
Glass bottle
Catalog number ACR139085000
Price (MYR)
565.00
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
565.00
Add to cart
Chemical Identifiers
CAS58-95-7
IUPAC Name2,5,7,8-tetramethyl-2-(4,8,12-trimethyltridecyl)-3,4-dihydro-2H-1-benzopyran-6-yl acetate
Molecular FormulaC31H52O3
InChI KeyZAKOWWREFLAJOT-UHFFFAOYNA-N
SMILESCC(C)CCCC(C)CCCC(C)CCCC1(C)CCC2=C(C)C(OC(C)=O)=C(C)C(C)=C2O1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=96.0 %
Appearance (Form)Viscous liquid
Refractive index1.4950 to 1.4990 (20°C, 589 nm)
Appearance (Color)Clear yellow