p-Toluenesulfonyl chloride, 99+%, Thermo Scientific Chemicals
p-Toluenesulfonyl chloride, 99+%, Thermo Scientific Chemicals
p-Toluenesulfonyl chloride, 99+%, Thermo Scientific Chemicals
p-Toluenesulfonyl chloride, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
p-Toluenesulfonyl chloride, 99+%, Thermo Scientific Chemicals
CAS: 98-59-9 | C7H7ClO2S | 190.64 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
2 kg
Packaging:
Glass bottle
Catalog number ACR139035000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Materialp-Toluenesulfonyl chloride
CAS98-59-9
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
May be corrosive to metals.
Causes skin irritation.
Causes serious eye damage.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre