p-Toluenesulfonic acid monohydrate, 99%, extra pure, Thermo Scientific Chemicals
p-Toluenesulfonic acid monohydrate, 99%, extra pure, Thermo Scientific Chemicals
p-Toluenesulfonic acid monohydrate, 99%, extra pure, Thermo Scientific Chemicals
p-Toluenesulfonic acid monohydrate, 99%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
p-Toluenesulfonic acid monohydrate, 99%, extra pure, Thermo Scientific Chemicals
CAS: 6192-52-5 | C7H10O4S | 190.213 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
500 g
2.5 kg
Packaging:
View all
Plastic bottle
Catalog number ACR139021000
Price (MYR)
427.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
427.00
Add to cart
Chemical Identifiers
CAS57467-17-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (Non-aqueous acid-base Titration)≥97.5 to ≤102.5%