p-Toluenesulfonic acid monohydrate, 99%, extra pure, Thermo Scientific Chemicals
p-Toluenesulfonic acid monohydrate, 99%, extra pure, Thermo Scientific Chemicals
p-Toluenesulfonic acid monohydrate, 99%, extra pure, Thermo Scientific Chemicals
p-Toluenesulfonic acid monohydrate, 99%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
p-Toluenesulfonic acid monohydrate, 99%, extra pure, Thermo Scientific Chemicals
CAS: 6192-52-5 | C7H10O4S | 190.213 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
View all
Glass Bottle
Catalog number ACR139020050
Price (MYR)
380.00
Add to cart
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
380.00
Add to cart
Chemical Identifiers
CAS993-16-8
Molecular FormulaCH9Cl3Sn
InChI KeyVLOZTKJSAQVQRJ-UHFFFAOYSA-N
SMILESCl.Cl.Cl.C[SnH3]
Molecular Weight (g/mol)246.14
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Proton NMRConforms to structure
FormPowder or crystals or crystalline powder
Assay from Supplier's CofA≥96.0%