p-Toluenesulfonic acid monohydrate, 99%, extra pure, Thermo Scientific Chemicals
p-Toluenesulfonic acid monohydrate, 99%, extra pure, Thermo Scientific Chemicals
p-Toluenesulfonic acid monohydrate, 99%, extra pure, Thermo Scientific Chemicals
p-Toluenesulfonic acid monohydrate, 99%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
p-Toluenesulfonic acid monohydrate, 99%, extra pure, Thermo Scientific Chemicals
CAS: 6192-52-5 | C7H10O4S | 190.213 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
5 g
100 g
500 g
Packaging:
View all
Plastic bottle
Catalog number ACR139020025
Price (MYR)
903.00
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
903.00
Add to cart
Chemical Identifiers
CAS5989-81-1
IUPAC Name(2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-{[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol hydrate
Molecular FormulaC12H24O12
InChI KeyWSVLPVUVIUVCRA-KPKNDVKVSA-N
SMILESO.OC[C@H]1O[C@@H](O[C@H]2[C@H](O)[C@@H](O)[C@@H](O)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=99.50 %
Water4.5 to 5.5 % (K.F.)
Specific optical rotationOn dry substance
Loss on drying=<0.5 % (80°C, 2 h)
AbsorbancePasses test