p-Toluenesulfinic acid, sodium salt hydrate, 98+%, Thermo Scientific Chemicals
p-Toluenesulfinic acid, sodium salt hydrate, 98+%, Thermo Scientific Chemicals
p-Toluenesulfinic acid, sodium salt hydrate, 98+%, Thermo Scientific Chemicals
p-Toluenesulfinic acid, sodium salt hydrate, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
p-Toluenesulfinic acid, sodium salt hydrate, 98+%, Thermo Scientific Chemicals
CAS: 207801-20-5 | C7H7NaO2S | 178.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Plastic bottle
Catalog number ACR139011000
Price (MYR)
653.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
653.00
Add to cart
Chemical Identifiers
CAS130-15-4
IUPAC Name1,4-dihydronaphthalene-1,4-dione
Molecular FormulaC10H6O2
InChI KeyFRASJONUBLZVQX-UHFFFAOYSA-N
SMILESO=C1C=CC(=O)C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Khaki
GC>=99 %
Appearance (Form)Powder
Water=<6 % (K.F.)