Thiourea, 99%, extra pure, Thermo Scientific Chemicals
Thiourea, 99%, extra pure, Thermo Scientific Chemicals
Thiourea, 99%, extra pure, Thermo Scientific Chemicals
Thiourea, 99%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Thiourea, 99%, extra pure, Thermo Scientific Chemicals
Thiourea, Thiocarbamide, CAS # 62-56-6, is an analogue of urea with a single sulfur-substituted oxygen.
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number ACR138915000
Price (MYR)
499.00
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
499.00
Add to cart
Chemical Identifiers
CAS5977-14-0
IUPAC Name3-oxobutanamide
Molecular FormulaC4H7NO2
InChI KeyGCPWJFKTWGFEHH-UHFFFAOYSA-N
SMILESCC(=O)CC(N)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point52°C to 58°C
HPLC>=96.0 %
General description
• Thiourea is a derivative of the biological compound, urea
• Thiourea is highly toxic when it decomposes and acts as a carcinogen
Applications
• Thiourea can be used as a denaturant in lysis buffers and similar sample buffers that require solubilization of proteins
• Thiourea can act as an antioxidant and chromophore
• Thiourea can be used in the study of cellular redox, autophagy, and aging
RUO – Research Use Only
General References:
  1. Wang, Y.; Jin, X.; He, L.; Zhang, W. Inhibitory effect of thiourea on biological nitrification process and its eliminating method. Water Sci Technol. 2017 75(12), 2900-2907.
  2. Park, S.; Lee, H.; Chae, J.B.; Kim, C. A Thiourea-Containing Fluorescent Chemosensor for Detecting Ga3. J Fluoresc. 2020 30(6), 1457-1462.