Thiosalicylic acid, 98%, Thermo Scientific Chemicals
Thiosalicylic acid, 98%, Thermo Scientific Chemicals
Thiosalicylic acid, 98%, Thermo Scientific Chemicals
Thiosalicylic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Thiosalicylic acid, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number ACR138891000
Price (MYR)
573.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
573.00
Add to cart
Chemical Identifiers
CAS59489-71-3
IUPAC Name5-bromopyrazin-2-amine
Molecular FormulaC4H4BrN3
InChI KeyKRRTXVSBTPCDOS-UHFFFAOYSA-N
SMILESNC1=CN=C(Br)C=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow to yellow
FormPowder
Assay (GC)≥96.0%
Melting Point (clear melt)111.0-117.0?C