2-Mercaptopropionic acid, 95%, Thermo Scientific Chemicals
2-Mercaptopropionic acid, 95%, Thermo Scientific Chemicals
2-Mercaptopropionic acid, 95%, Thermo Scientific Chemicals
2-Mercaptopropionic acid, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Mercaptopropionic acid, 95%, Thermo Scientific Chemicals
CAS: 79-42-5 | C3H6O2S | 106.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Packaging:
Glass bottle
Catalog number ACR138795000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2-Mercaptopropionic acid
Name Note95%
CAS79-42-5
CAS Min %94.0
Health Hazard 1GHS Signal Word: Danger