2-Mercaptopropionic acid, 95%, Thermo Scientific Chemicals
2-Mercaptopropionic acid, 95%, Thermo Scientific Chemicals
2-Mercaptopropionic acid, 95%, Thermo Scientific Chemicals
2-Mercaptopropionic acid, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Mercaptopropionic acid, 95%, Thermo Scientific Chemicals
CAS: 79-42-5 | C3H6O2S | 106.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Catalog number ACR138791000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS13280-60-9
IUPAC Name5-amino-2-nitrobenzoic acid
Molecular FormulaC7H6N2O4
InChI KeyKZZWQCKYLNIOBT-UHFFFAOYSA-N
SMILESNC1=CC=C(C(=C1)C(O)=O)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to brown
FormCrystals or powder or crystalline powder
Assay (HPLC)≥94.0%
Assay (Aqueous acid-base Titration)≥94.0 to ≤106.0%