Thioacetic acid, 98%, Thermo Scientific Chemicals
Thioacetic acid, 98%, Thermo Scientific Chemicals
Thioacetic acid, 98%, Thermo Scientific Chemicals
Thioacetic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Thioacetic acid, 98%, Thermo Scientific Chemicals
CAS: 507-09-5 | C2H4OS | 76.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Glass bottle
Catalog number ACR138785000
Price (MYR)
1,395.00
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,395.00
Add to cart
Chemical Identifiers
CAS12013-82-0
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities1% max.