Thioacetic acid, 98%, Thermo Scientific Chemicals
Thioacetic acid, 98%, Thermo Scientific Chemicals
Thioacetic acid, 98%, Thermo Scientific Chemicals
Thioacetic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Thioacetic acid, 98%, Thermo Scientific Chemicals
CAS: 507-09-5 | C2H4OS | 76.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number ACR138781000
Price (MYR)
602.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
602.00
Add to cart
Chemical Identifiers
CAS12027-06-4
IUPAC Nameammonium iodide
Molecular FormulaH4IN
InChI KeyUKFWSNCTAHXBQN-UHFFFAOYSA-N
SMILES[NH4+].[I-]
View more
SpecificationsSpecification SheetSpecification Sheet
pH4.5 to 6.5 (5 % at 20°C)
Chloride and Bromide=<0.05 % (as Cl)
Phosphate (PO4)=<10 ppm
Calcium (Ca)=<20 ppm
Appearance (Form)Crystalline powder