DL-Thioctic Acid, 98+%
DL-Thioctic Acid, 98+%
DL-Thioctic Acid, 98+%
DL-Thioctic Acid, 98+%
Thermo Scientific Chemicals
DL-Thioctic Acid, 98+%
CAS: 1077-28-7 | C8H14O2S2 | 206.32 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number ACR138721000
Price (MYR)
6,125.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
6,125.00
Add to cart
Chemical Identifiers
CAS4021-11-8
IUPAC Name2-methylpyridine-4-carboxylic acid
Molecular FormulaC7H7NO2
InChI KeyPMDHIMMPXRSDML-UHFFFAOYSA-N
SMILESCC1=NC=CC(=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to brown
Infrared spectrumConforms
Appearance (Form)Crystalline powder or solid
HPLC>=96.0 %