DL-Thioctic Acid, 98+%
DL-Thioctic Acid, 98+%
DL-Thioctic Acid, 98+%
DL-Thioctic Acid, 98+%
Thermo Scientific Chemicals
DL-Thioctic Acid, 98+%
CAS: 1077-28-7 | C8H14O2S2 | 206.32 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Packaging:
Glass bottle
Catalog number ACR138720250
Price (MYR)
2,204.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,204.00
Add to cart
Chemical Identifiers
CAS654058-04-5
IUPAC Name1,3-dimethyl-1H-imidazol-3-ium dimethyl phosphate
Molecular FormulaC7H15N2O4P
InChI KeyGSGLHYXFTXGIAQ-UHFFFAOYSA-M
SMILESCOP([O-])(=O)OC.CN1C=C[N+](C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid or viscous liquid
Identification (FTIR)Conforms
Refractive Index1.4820-1.4860 @ 20?C
Appearance (Color)Clear pale yellow to yellow
Assay (Non-aqueous acid-base Titration)≥97.5 to ≤102.5%