p-Toluenethiol, 98%, Thermo Scientific Chemicals
p-Toluenethiol, 98%, Thermo Scientific Chemicals
p-Toluenethiol, 98%, Thermo Scientific Chemicals
p-Toluenethiol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
p-Toluenethiol, 98%, Thermo Scientific Chemicals
CAS: 106-45-6 | C7H8S | 124.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number ACR138715000
Price (MYR)
1,668.00
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,668.00
Add to cart
Chemical Identifiers
CAS79-41-4
IUPAC Name2-methylprop-2-enoic acid
Molecular FormulaC4H6O2
InChI KeyCERQOIWHTDAKMF-UHFFFAOYSA-N
SMILESCC(=C)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=99.4 %
Color scale=<20 APHA
Appearance (Form)Clear liquid
Refractive index1.4295 to 1.4315 (20°C, 589 nm)