Thiocarbanilide, 98%, Thermo Scientific Chemicals
Thiocarbanilide, 98%, Thermo Scientific Chemicals
Thiocarbanilide, 98%, Thermo Scientific Chemicals
Thiocarbanilide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Thiocarbanilide, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Catalog number ACR138701000
Price (MYR)
408.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
408.00
Add to cart
Chemical Identifiers
CAS6020-87-7
IUPAC Name2-(N-methylcarbamimidamido)acetic acid hydrate
Molecular FormulaC4H11N3O3
InChI KeyMEJYXFHCRXAUIL-UHFFFAOYSA-N
SMILESO.CN(CC(O)=O)C(N)=N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
Infrared spectrumConforms
Residue after ignition=<1 %
HPLC>=98.5 % (on dry substance)
Heavy metals (as Pb)=<10 ppm