Tetramethylene sulfoxide, 97%, Thermo Scientific Chemicals
Tetramethylene sulfoxide, 97%, Thermo Scientific Chemicals
Tetramethylene sulfoxide, 97%, Thermo Scientific Chemicals
Tetramethylene sulfoxide, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Tetramethylene sulfoxide, 97%, Thermo Scientific Chemicals
CAS: 1600-44-8 | C4H8OS | 104.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number ACR138441000
Price (MYR)
2,651.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,651.00
Add to cart
Chemical Identifiers
CAS621-84-1
SpecificationsSpecification SheetSpecification Sheet
Water Content (Karl Fischer Titration)≤0.75%
Assay (GC)≥98.5%
Melting Point (clear melt)83-91?C
Appearance (Color)White to pale cream
FormCrystalline powder or flakes