Tetramethylene sulfoxide, 97%, Thermo Scientific Chemicals
Tetramethylene sulfoxide, 97%, Thermo Scientific Chemicals
Tetramethylene sulfoxide, 97%, Thermo Scientific Chemicals
Tetramethylene sulfoxide, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Tetramethylene sulfoxide, 97%, Thermo Scientific Chemicals
CAS: 1600-44-8 | C4H8OS | 104.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number ACR138440250
Price (MYR)
888.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
888.00
Add to cart
Chemical Identifiers
CAS142-96-1
IUPAC Name1-butoxybutane
Molecular FormulaC8H18O
InChI KeyDURPTKYDGMDSBL-UHFFFAOYSA-N
SMILESCCCCOCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<0.005 %
Residue after evaporation=<0.002 %
GC>=99.0 %
Appearance (Color)Clear colorless
Appearance (Form)Liquid