1,2,4,5-Tetramethylbenzene, 97+%, Thermo Scientific Chemicals
1,2,4,5-Tetramethylbenzene, 97+%, Thermo Scientific Chemicals
1,2,4,5-Tetramethylbenzene, 97+%, Thermo Scientific Chemicals
1,2,4,5-Tetramethylbenzene, 97+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,2,4,5-Tetramethylbenzene, 97+%, Thermo Scientific Chemicals
CAS: 95-93-2 | C10H14 | 134.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Catalog number ACR138370050
Price (MYR)
331.00
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
331.00
Add to cart
Specifications
Chemical Name or Material1,2,4,5-Tetramethylbenzene
CAS95-93-2
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Flammable solid.
Very toxic to aquatic life with long lasting effects.
Health Hazard 3GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid release to the environment.