1,2,3,4-Tetrahydroisoquinoline, 95%, Thermo Scientific Chemicals
1,2,3,4-Tetrahydroisoquinoline, 95%, Thermo Scientific Chemicals
1,2,3,4-Tetrahydroisoquinoline, 95%, Thermo Scientific Chemicals
1,2,3,4-Tetrahydroisoquinoline, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,2,3,4-Tetrahydroisoquinoline, 95%, Thermo Scientific Chemicals
CAS: 91-21-4 | C9H11N | 133.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number ACR138200250
Price (MYR)
637.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
637.00
Add to cart
Chemical Identifiers
CAS85068-32-2
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥96.0%
Refractive Index1.4205-1.4255 @ 20?C