Tetrahydrofurfuryl alcohol, 98%, Thermo Scientific Chemicals
Tetrahydrofurfuryl alcohol, 98%, Thermo Scientific Chemicals
Tetrahydrofurfuryl alcohol, 98%, Thermo Scientific Chemicals
Tetrahydrofurfuryl alcohol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Tetrahydrofurfuryl alcohol, 98%, Thermo Scientific Chemicals
CAS: 97-99-4 | C5H10O2 | 102.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
Packaging:
Glass bottle
Catalog number ACR138180025
Price (MYR)
742.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
742.00
Add to cart
Specifications
Chemical Name or MaterialTetrahydrofurfuryl alcohol
Melting Point-80.0°C
Name Note98%
CAS97-99-4
Health Hazard 1GHS Signal Word: Danger