DL-Tartaric acid, 99.5%, Thermo Scientific Chemicals
DL-Tartaric acid, 99.5%, Thermo Scientific Chemicals
DL-Tartaric acid, 99.5%, Thermo Scientific Chemicals
DL-Tartaric acid, 99.5%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
DL-Tartaric acid, 99.5%, Thermo Scientific Chemicals
CAS: 133-37-9 | C4H6O6 | 150.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number ACR137871000
Price (MYR)
408.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
408.00
Add to cart
Chemical Identifiers
CAS552-30-7
IUPAC Name1,3-dioxo-1,3-dihydro-2-benzofuran-5-carboxylic acid
Molecular FormulaC9H4O5
InChI KeySRPWOOOHEPICQU-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=C2C(=O)OC(=O)C2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or flakes
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
Appearance (Color)White to cream
Identification (FTIR)Conforms
Melting Point161-171?C