DL-Tartaric acid, 99.5%, Thermo Scientific Chemicals
DL-Tartaric acid, 99.5%, Thermo Scientific Chemicals
DL-Tartaric acid, 99.5%, Thermo Scientific Chemicals
DL-Tartaric acid, 99.5%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
DL-Tartaric acid, 99.5%, Thermo Scientific Chemicals
CAS: 133-37-9 | C4H6O6 | 150.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
100 g
500 g
Packaging:
Plastic bottle
Catalog number ACR137870025
Price (MYR)
2,411.00
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
2,411.00
Add to cart
Chemical Identifiers
CAS106-91-2
IUPAC Name(oxiran-2-yl)methyl 2-methylprop-2-enoate
Molecular FormulaC7H10O3
InChI KeyVOZRXNHHFUQHIL-UHFFFAOYNA-N
SMILESCC(=C)C(=O)OCC1CO1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Water=<0.1 % (K.F.)
Refractive index1.4490 to 1.4510 (20°C, 589 nm)
Appearance (Form)Clear liquid
Color scale=<30 APHA