L(+)-Tartaric acid, 99+%, Thermo Scientific Chemicals
L(+)-Tartaric acid, 99+%, Thermo Scientific Chemicals
L(+)-Tartaric acid, 99+%, Thermo Scientific Chemicals
L(+)-Tartaric acid, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
L(+)-Tartaric acid, 99+%, Thermo Scientific Chemicals
CAS: 87-69-4 | C4H6O6 | 150.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number ACR137855000
Price (MYR)
501.00
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
501.00
Add to cart
Chemical Identifiers
CAS589-98-0
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4240-1.4290 @ 20?C
Appearance (Color)Clear colorless
Assay (GC)≥96.0%