L(+)-Tartaric acid, 99+%, Thermo Scientific Chemicals
L(+)-Tartaric acid, 99+%, Thermo Scientific Chemicals
L(+)-Tartaric acid, 99+%, Thermo Scientific Chemicals
L(+)-Tartaric acid, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
L(+)-Tartaric acid, 99+%, Thermo Scientific Chemicals
CAS: 87-69-4 | C4H6O6 | 150.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
500 g
Packaging:
Plastic bottle
Catalog number ACR137850025
Price (MYR)
1,062.00
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,062.00
Add to cart
Chemical Identifiers
CAS6153-39-5
IUPAC Name5-methylbenzene-1,3-diol hydrate
Molecular FormulaC7H10O3
InChI KeyNBKPNAMTHBIMLA-UHFFFAOYSA-N
SMILESO.CC1=CC(O)=CC(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
FormCrystals or crystalline powder
Water Content (Karl Fischer Titration)10-15%
Appearance (Color)White to pale brown to pink-brown