3'-Chloroacetophenone, 98%, Thermo Scientific Chemicals
3'-Chloroacetophenone, 98%, Thermo Scientific Chemicals
3'-Chloroacetophenone, 98%, Thermo Scientific Chemicals
3'-Chloroacetophenone, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3'-Chloroacetophenone, 98%, Thermo Scientific Chemicals
CAS: 99-02-5 | C8H7ClO | 154.59 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number ACR134840250
Price (MYR)
802.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
802.00
Add to cart
Chemical Identifiers
CAS7789-17-5
IUPAC Namecaesium(1+) iodide
Molecular FormulaCsI
InChI KeyXQPRBTXUXXVTKB-UHFFFAOYSA-M
SMILES[I-].[Cs+]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.001% max.