Potassium hydroxide, ca. 85%, for analysis, pellets, Thermo Scientific Chemicals
Potassium hydroxide, ca. 85%, for analysis, pellets, Thermo Scientific Chemicals
Potassium hydroxide, ca. 85%, for analysis, pellets, Thermo Scientific Chemicals
Potassium hydroxide, ca. 85%, for analysis, pellets, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Potassium hydroxide, ca. 85%, for analysis, pellets, Thermo Scientific Chemicals
CAS: 1310-58-3 | HKO | 56.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
Catalog number ACR134062500
Price (MYR)
425.00
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
425.00
Add to cart
Chemical Identifiers
CAS24424-99-5
IUPAC Namedi-tert-butyl dicarbonate
Molecular FormulaC10H18O5
InChI KeyDYHSDKLCOJIUFX-UHFFFAOYSA-N
SMILESCC(C)(C)OC(=O)OC(=O)OC(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless or white to pale yellow
FormCrystals or powder or crystalline powder or fused/lumpy solid or liquid as melt
Assay from Supplier's CofA≥97.0%