Potassium hydroxide, ca. 85%, for analysis, pellets, Thermo Scientific Chemicals
Potassium hydroxide, ca. 85%, for analysis, pellets, Thermo Scientific Chemicals
Potassium hydroxide, ca. 85%, for analysis, pellets, Thermo Scientific Chemicals
Potassium hydroxide, ca. 85%, for analysis, pellets, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Potassium hydroxide, ca. 85%, for analysis, pellets, Thermo Scientific Chemicals
CAS: 1310-58-3 | HKO | 56.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
Packaging:
Plastic bottle
Catalog number ACR134060010
Price (MYR)
721.00
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
721.00
Add to cart
Chemical Identifiers
CAS3510-66-5
IUPAC Name2-bromo-5-methylpyridine
Molecular FormulaC6H6BrN
InChI KeyYWNJQQNBJQUKME-UHFFFAOYSA-N
SMILESCC1=CC=C(Br)N=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)40-47°C
Appearance (Color)White to yellow to pale brown
FormCrystals or powder or crystalline powder or low melting crystalline solid
Assay (GC)≥98.0%