Ammonia, ca. 7N solution in methanol, Thermo Scientific Chemicals
Ammonia, ca. 7N solution in methanol, Thermo Scientific Chemicals
Ammonia, ca. 7N solution in methanol, Thermo Scientific Chemicals
Ammonia, ca. 7N solution in methanol, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ammonia, ca. 7N solution in methanol, Thermo Scientific Chemicals
Ammonia, H3N, CAS Number-7664-41-7, 67-56-1, am-fol, ammonia, ammonia anhydrous, ammonia, anhydrous, ammonia gas, ammoniak, ammoniakgas, anhydrous ammonia, nitro-sil, spirit of hartshorn, 2.5L, CHEBI:16134, Colorless to Yellow, 6.0 to 7.5N, 0.7700g/mL, 05,15, 14 deg.C, 17.03, GHS Signal Word: Danger
Have Questions?
Catalog NumberQuantityPackaging
ACR1337100252.5 LGlass Bottle
Catalog number ACR133710025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS9002-18-0
IUPAC Name2-({4-hydroxy-3-methyl-2,6-dioxabicyclo[3.2.1]octan-8-yl}oxy)-6-(hydroxymethyl)-4-methoxyoxane-3,5-diol
Molecular FormulaC14H24O9
InChI KeyGYYDPBCUIJTIBM-UHFFFAOYNA-N
SMILESCOC1C(O)C(CO)OC(OC2C3COC2C(O)C(C)O3)C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
pH6.0 to 7.5 (10 % suspension)
Appearance (Color)White to cream to light yellow to beige
Ash=<5 %
Solubility(1.5 % in hot water) Clear to slightly