Hydrogen chloride, pure, 5 to 6N solution in 2-propanol, Thermo Scientific Chemicals
Hydrogen chloride, pure, 5 to 6N solution in 2-propanol, Thermo Scientific Chemicals
Hydrogen chloride, pure, 5 to 6N solution in 2-propanol, Thermo Scientific Chemicals
Hydrogen chloride, pure, 5 to 6N solution in 2-propanol, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Hydrogen chloride, pure, 5 to 6N solution in 2-propanol, Thermo Scientific Chemicals
CAS: 7647-01-0 | ClH | 36.46 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Packaging:
Plastic bottle
Catalog number ACR133700010
Price (MYR)
848.00
Add to cart
Quantity:
1 L
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
848.00
Add to cart
Chemical Identifiers
CAS9048-46-8
MDL NumberMFCD00145743
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow with tan to pale green
Assay (special)Endotoxin: ó 3 EU/mg (LAL)
Loss on Dryingó 5.0%
Protease activityó 0.005 U/ mg (enzymatic assay)
FormLyophilized powder