1,1,2,2-Tetrachloroethane-d2 99 atom% D, Thermo Scientific Chemicals
1,1,2,2-Tetrachloroethane-d2 99 atom% D, Thermo Scientific Chemicals
1,1,2,2-Tetrachloroethane-d2 99 atom% D, Thermo Scientific Chemicals
1,1,2,2-Tetrachloroethane-d2 99 atom% D, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,1,2,2-Tetrachloroethane-d2 99 atom% D, Thermo Scientific Chemicals
CAS: 33685-54-0 | C2H2Cl4 | 169.85 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR13358020020 mLGlass Bottle
Catalog number ACR133580200
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
20 mL
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS693-02-7
IUPAC Namehex-1-yne
Molecular FormulaC6H10
InChI KeyCGHIBGNXEGJPQZ-UHFFFAOYSA-N
SMILESCCCCC#C
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Refractive Index1.3970-1.4010 @ 20?C
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥98.0%
FormLiquid
For NMR
RUO – Research Use Only