5-Sulfosalicylic Acid Dihydrate, 98%, Thermo Scientific Chemicals
5-Sulfosalicylic Acid Dihydrate, 98%, Thermo Scientific Chemicals
5-Sulfosalicylic Acid Dihydrate, 98%, Thermo Scientific Chemicals
5-Sulfosalicylic Acid Dihydrate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
5-Sulfosalicylic Acid Dihydrate, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Plastic bottle
Catalog number ACR132862500
Price (MYR)
529.00
BOT
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
529.00
BOT
Add to cart
Chemical Identifiers
CAS108354-78-5
IUPAC Namemethyl 2-amino-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylate
Molecular FormulaC10H13NO2S
InChI KeyDKYYKIHEIOOWRB-UHFFFAOYSA-N
SMILESCOC(=O)C1=C(N)SC2=C1CCCC2
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
FormCrystals or powder or crystalline powder
Appearance (Color)Pale cream to cream to yellow
Assay (GC)≥97.5%
Melting Point (clear melt)128.0-134.0?C
Indicator
RUO – Research Use Only