5-Sulfosalicylic Acid Dihydrate, 98%, Thermo Scientific Chemicals
5-Sulfosalicylic Acid Dihydrate, 98%, Thermo Scientific Chemicals
5-Sulfosalicylic Acid Dihydrate, 98%, Thermo Scientific Chemicals
5-Sulfosalicylic Acid Dihydrate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
5-Sulfosalicylic Acid Dihydrate, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
2.5 kg
Packaging:
Plastic bottle
Catalog number ACR132860010
Price (MYR)
923.00
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
923.00
Add to cart
Chemical Identifiers
CAS5965-83-3, 97-05-2
Specifications
Chemical Name or Material5-Sulfosalicylic acid dihydrate
CAS5965-83-3
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0
View more
Indicator
RUO – Research Use Only