Stigmasterol, 95%, Thermo Scientific Chemicals
Stigmasterol, 95%, Thermo Scientific Chemicals
Stigmasterol, 95%, Thermo Scientific Chemicals
Stigmasterol, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Stigmasterol, 95%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number ACR132760050
Price (MYR)
1,162.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,162.00
Add to cart
Specifications
Chemical Name or MaterialStigmasterol
Name Note95%
CAS83-48-7
Melting Point161.0°C to 170.0°C