beta-Sitosterol, with ca. 10% campesterol, ca. 75% beta-sitosterol
beta-Sitosterol, with ca. 10% campesterol, ca. 75% beta-sitosterol
beta-Sitosterol, with ca. 10% campesterol, ca. 75% beta-sitosterol
beta-Sitosterol, with ca. 10% campesterol, ca. 75% beta-sitosterol
Thermo Scientific Chemicals
beta-Sitosterol, with ca. 10% campesterol, ca. 75% beta-sitosterol
CAS: 83-46-5 | C29H50O | 414.718 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR132725000500 gPlastic bottle
Catalog number ACR132725000
Price (MYR)
1,872.00
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,872.00
Add to cart
Chemical Identifiers
CAS883502-07-6
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%