beta-Sitosterol, with ca. 10% campesterol, ca. 75% beta-sitosterol, Thermo Scientific Chemicals
beta-Sitosterol, with ca. 10% campesterol, ca. 75% beta-sitosterol, Thermo Scientific Chemicals
beta-Sitosterol, with ca. 10% campesterol, ca. 75% beta-sitosterol, Thermo Scientific Chemicals
beta-Sitosterol, with ca. 10% campesterol, ca. 75% beta-sitosterol, Thermo Scientific Chemicals
Thermo Scientific Chemicals
beta-Sitosterol, with ca. 10% campesterol, ca. 75% beta-sitosterol, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
Catalog number ACR132720050
Price (MYR)
479.00
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
479.00
Add to cart
Specifications
Chemical Name or Materialβ-Sitosterol
CAS83-46-5
ColorWhite
Loss on Drying3.5% max.
Infrared SpectrumAuthentic