Sesamol, 98%, Thermo Scientific Chemicals
Sesamol, 98%, Thermo Scientific Chemicals
Sesamol, 98%, Thermo Scientific Chemicals
Sesamol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Sesamol, 98%, Thermo Scientific Chemicals
CAS: 533-31-3 | C7H6O3 | 138.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number ACR132690050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS513-77-9
IUPAC Namebarium(2+) carbonate
Molecular FormulaCBaO3
InChI KeyAYJRCSIUFZENHW-UHFFFAOYSA-L
SMILES[Ba++].[O-]C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Impurity contentNa 0.02%
Impurity contentOxidizing substances (as NO₃) 0.005%
Impurity contentS 0.001%
Impurity contentSr 0.7%
Trace Metals(as Pb) 0.001%