Pyridine, 99+%, extra pure, Thermo Scientific Chemicals
Pyridine, 99+%, extra pure, Thermo Scientific Chemicals
Pyridine, 99+%, extra pure, Thermo Scientific Chemicals
Pyridine, 99+%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Pyridine, 99+%, extra pure, Thermo Scientific Chemicals
CAS: 110-86-1 | C5H5N | 79.102 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
1 L
2.5 L
Packaging:
View all
Glass Bottle
Catalog number ACR131780500
Price (MYR)
343.00
Add to cart
Quantity:
50 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
343.00
Add to cart
Chemical Identifiers
CAS144-55-8
IUPAC Namesodium hydrogen carbonate
Molecular FormulaCHNaO3
InChI KeyUIIMBOGNXHQVGW-UHFFFAOYSA-M
SMILES[Na+].OC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with HCl99.7 to 100.3 % (On dried substance)
Heavy metals (as Pb)=<5 ppm
Appearance (Color)White
Appearance (Form)Powder
Sulfur compounds (as SO4)=<0.003 %