Pyridine, 99+%, extra pure, Thermo Scientific Chemicals
Pyridine, 99+%, extra pure, Thermo Scientific Chemicals
Pyridine, 99+%, extra pure, Thermo Scientific Chemicals
Pyridine, 99+%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Pyridine, 99+%, extra pure, Thermo Scientific Chemicals
CAS: 110-86-1 | C5H5N | 79.102 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
50 mL
1 L
Packaging:
View all
Glass Bottle
Catalog number ACR131780025
Price (MYR)
1,519.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
1,519.00
Add to cart
Chemical Identifiers
CAS16853-85-3
IUPAC Namelithium(1+) alumanuide
Molecular FormulaAlH4Li
InChI KeyOCZDCIYGECBNKL-UHFFFAOYSA-N
SMILES[Li+].[AlH4-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solution
Strength9.5 to 10.5 % (as LiAlH4)
AppearanceMay develop some turbidity or precipitate
Appearance (Color)Colorless to grey