Pyridine, 99+%, extra pure, Thermo Scientific Chemicals
Pyridine, 99+%, extra pure, Thermo Scientific Chemicals
Pyridine, 99+%, extra pure, Thermo Scientific Chemicals
Pyridine, 99+%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Pyridine, 99+%, extra pure, Thermo Scientific Chemicals
CAS: 110-86-1 | C5H5N | 79.102 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
50 mL
2.5 L
Packaging:
View all
EcoSafPak™ Glass Bottle
Catalog number ACR131780010
Price (MYR)
797.00
Add to cart
Quantity:
1 L
Packaging:
EcoSafPak™ Glass Bottle
Request bulk or custom format
Price (MYR)
797.00
Add to cart
Chemical Identifiers
CAS7789-68-6
IUPAC Nametitanium(4+) tetrabromide
Molecular FormulaBr4Ti
InChI KeyUBZYKBZMAMTNKW-UHFFFAOYSA-J
SMILES[Ti+4].[Br-].[Br-].[Br-].[Br-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Orange
Titration Argentometric97.5 to 102.5 %
Appearance (Form)Powder and/or lumps or low melting solid