Pyrazole, 98%, pure, Thermo Scientific Chemicals
Pyrazole, 98%, pure, Thermo Scientific Chemicals
Pyrazole, 98%, pure, Thermo Scientific Chemicals
Pyrazole, 98%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Pyrazole, 98%, pure, Thermo Scientific Chemicals
CAS: 288-13-1 | C3H4N2 | 68.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number ACR131741000
Price (MYR)
875.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
875.00
Add to cart
Specifications
Chemical Name or MaterialPyrazole
Name Note98%
CAS288-13-1
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement: