2-Pyrazinecarboxylic acid, 99%, Thermo Scientific Chemicals
2-Pyrazinecarboxylic acid, 99%, Thermo Scientific Chemicals
2-Pyrazinecarboxylic acid, 99%, Thermo Scientific Chemicals
2-Pyrazinecarboxylic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Pyrazinecarboxylic acid, 99%, Thermo Scientific Chemicals
CAS: 98-97-5 | C5H4N2O2 | 124.1 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Plastic bottle
Catalog number ACR131711000
Price (MYR)
994.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
994.00
Add to cart
Chemical Identifiers
CAS616-30-8
IUPAC Name3-aminopropane-1,2-diol
Molecular FormulaC3H9NO2
InChI KeyKQIGMPWTAHJUMN-UHFFFAOYNA-N
SMILESNCC(O)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with HClO4>=97.5 %
Water=<0.5 % (K.F.)
Refractive index1.4900 to 1.4940 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Viscous liquid