2-Pyrazinecarboxylic acid, 99%, Thermo Scientific Chemicals
2-Pyrazinecarboxylic acid, 99%, Thermo Scientific Chemicals
2-Pyrazinecarboxylic acid, 99%, Thermo Scientific Chemicals
2-Pyrazinecarboxylic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Pyrazinecarboxylic acid, 99%, Thermo Scientific Chemicals
CAS: 98-97-5 | C5H4N2O2 | 124.1 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Plastic bottle
Catalog number ACR131710250
Price (MYR)
482.00
Add to cart
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
482.00
Add to cart
Chemical Identifiers
CAS3680-71-5
IUPAC Name1H,4H,7H-pyrrolo[2,3-d]pyrimidin-4-one
Molecular FormulaC6H5N3O
InChI KeyFBMZEITWVNHWJW-UHFFFAOYSA-N
SMILESO=C1N=CNC2=C1C=CN2
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>96.0%