Propiolic Acid, 98%, Thermo Scientific Chemicals
Propiolic Acid, 98%, Thermo Scientific Chemicals
Propiolic Acid, 98%, Thermo Scientific Chemicals
Propiolic Acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Propiolic Acid, 98%, Thermo Scientific Chemicals
CAS: 471-25-0 | C3H2O2 | 70.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
10 mL
Packaging:
Glass bottle
Catalog number ACR131500250
Price (MYR)
1,036.00
Add to cart
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,036.00
Add to cart
Specifications
Chemical Name or MaterialPropiolic acid
CAS471-25-0
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Flammable liquid and vapor.
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Health Hazard 3GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Gentl