Propargylamine, 99%, Thermo Scientific Chemicals
Propargylamine, 99%, Thermo Scientific Chemicals
Propargylamine, 99%, Thermo Scientific Chemicals
Propargylamine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Propargylamine, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Catalog number ACR131460050
Price (MYR)
848.00
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
848.00
Add to cart
Specifications
Chemical Name or MaterialPropargylamine
Name Note99%
CAS2450-71-7
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Highly flammable liquid and vapor.
Harmful if swallowed.
Toxic in contact with skin.