Propargyl alcohol, 99%, Thermo Scientific Chemicals
Propargyl alcohol, 99%, Thermo Scientific Chemicals
Propargyl alcohol, 99%, Thermo Scientific Chemicals
Propargyl alcohol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Propargyl alcohol, 99%, Thermo Scientific Chemicals
CAS: 107-19-7 | C3H4O | 56.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
500 mL
Packaging:
Glass bottle
Catalog number ACR131450010
Price (MYR)
816.00
Add to cart
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
816.00
Add to cart
Specifications
Chemical Name or MaterialPropargyl alcohol
Name Note99%
CAS107-19-7
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Toxic to aquatic life with long lasting effects.
Toxic if swallowed.
Fatal if inhaled.
Fatal in contact with skin.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Suspected o