1,3-Propane sultone, 97%, Thermo Scientific Chemicals
1,3-Propane sultone, 97%, Thermo Scientific Chemicals
1,3-Propane sultone, 97%, Thermo Scientific Chemicals
1,3-Propane sultone, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,3-Propane sultone, 97%, Thermo Scientific Chemicals
CAS: 1120-71-4 | C3H6O3S | 122.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
Catalog number ACR131421000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Chemical Identifiers
CAS1987-50-4
IUPAC Name4-heptylphenol
Molecular FormulaC13H20O
InChI KeyKNDDEFBFJLKPFE-UHFFFAOYSA-N
SMILESCCCCCCCC1=CC=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream to cream to brown
Identification (FTIR)Conforms
FormFused solid
Refractive Index1.5015-1.5055 @ 20?C
Melting Point (clear melt)26.0-32.0?C