1,3-Propane sultone, 97%, Thermo Scientific Chemicals
1,3-Propane sultone, 97%, Thermo Scientific Chemicals
1,3-Propane sultone, 97%, Thermo Scientific Chemicals
1,3-Propane sultone, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,3-Propane sultone, 97%, Thermo Scientific Chemicals
CAS: 1120-71-4 | C3H6O3S | 122.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
1 kg
Catalog number ACR131420250
Price (MYR)
611.00
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
611.00
Add to cart
Chemical Identifiers
CAS94-36-0
IUPAC Namebenzoyl benzenecarboperoxoate
Molecular FormulaC14H10O4
InChI KeyOMPJBNCRMGITSC-UHFFFAOYSA-N
SMILESO=C(OOC(=O)C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Moist granular powder
Appearance (Color)White
Impurity=<0.25 % Inorganic and organic hydrolizable chloride
Titration Iodometric73 to 77 %