Piperonyl alcohol, 98%, Thermo Scientific Chemicals
Piperonyl alcohol, 98%, Thermo Scientific Chemicals
Piperonyl alcohol, 98%, Thermo Scientific Chemicals
Piperonyl alcohol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Piperonyl alcohol, 98%, Thermo Scientific Chemicals
CAS: 495-76-1 | C8H8O3 | 152.15 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR13135025025 gGlass bottle
Catalog number ACR131350250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialPiperonyl alcohol
CAS495-76-1
Melting Point51.0°C to 55.0°C
ColorWhite
EINECS Number207-808-4